CAS RN 160068-88-2
CAS RN 160068-88-2
IUPAC Name:
4-(1,3,2-dioxaborinan-2-yl)benzaldehyde
Synonyms:
4-(1,3,2-dioxaborinan-2-yl)benzaldehyde
Molecular Weight (g/mol):
190.01
Molecular Formula:
C10H11BO3
InChi Key:
AMQBUMNOSGJNQL-UHFFFAOYSA-N
SMILES:
O=CC1=CC=C(C=C1)B1OCCCO1
1
–
1
of
1
results
1
Thermo Scientific Maybridge 4-(1,3,2-Dioxaborinan-2-yl)benzaldehyde, ≥97%, Thermo Scientific™
CAS: 160068-88-2 Molecular Formula: C10H11BO3 Molecular Weight (g/mol): 190.005 MDL Number: MFCD04039026 InChI Key: AMQBUMNOSGJNQL-UHFFFAOYSA-N Synonym: 4-1,3,2-dioxaborinan-2-yl benzaldehyde,4-formylbenzeneboronic acid, propane-1,3-diol cyclic ester,4-formylbenzeneboronic acid,propane-1,3-diol cyclic ester,4-1,3,2-dioxaboran-2-yl benzaldehyde,2-4-formylphenyl-1,3,2-dioxaborinane PubChem CID: 2794711 IUPAC Name: 4-(1,3,2-dioxaborinan-2-yl)benzaldehyde SMILES: B1(OCCCO1)C2=CC=C(C=C2)C=O