accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 151837-09-1

CH3NHH3CH3CONNNNNH2HOOPSOO

CAS RN 151837-09-1

IUPAC Name: triethylazanium 6-(6-amino-9H-purin-9-yl)-7-hydroxy-2-sulfanylidene-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate
Synonyms: TEAH 6-(6-aminopurin-9-yl)-7-hydroxy-2-sulfanylidene-tetrahydro-4H-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate
Molecular Weight (g/mol): 446.46
Molecular Formula: C16H27N6O5PS
InChi Key: OXIPZMKSNMRTIV-UHFFFAOYNA-N
SMILES: CC[NH+](CC)CC.NC1=C2N=CN(C3OC4COP([O-])(=S)OC4C3O)C2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.