CAS RN 146368-14-1
CAS RN 146368-14-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Tocris Bioscience™ Cyanine 5, SE
Red fluorescent dye (Reactive group: NHS ester). Application: confocal microscopy, fluorescence microscopy, flow cytometry
2
MedChemExpress CY5-SE
MedChemExpress Cy5-SE (Cy5 NHS Ester) is a reactive dye for the labeling of amino-groups in peptides, proteins, and oligonucleotides. This dye requires small amount of organic co-solvent (such as DMF or DMSO) to be used in labeling reaction.

3
Tocris Bioscience™ Cyanine 5, SE
Red fluorescent dye for the labeling of amines; supplied as NHS ester