missing translation for 'onlineSavingsMsg'
Få mere at vide

MedChemExpress CY5-SE

Artikelnummer. 16408768
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
16408768 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16408768 Leverandør MedChemExpress Leverandørnr. HYD0819

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Cy5-SE (Cy5 NHS Ester) is a reactive dye for the labeling of amino-groups in peptides, proteins, and oligonucleotides. This dye requires small amount of organic co-solvent (such as DMF or DMSO) to be used in labeling reaction.

This reagent is ideal for very cost-efficient labeling of soluble proteins, as well as all kinds of peptides and oligonucleotides. This reagent also works well in organic solvents for small molecule labeling. Excitation (nm):649, Emission (nm): 670.

Tekniske data

Mængde 10 mg
Produkttype Dye Reagents
Molekylvægt (g/mol) 753.88 g/mol
Opløselighed DMSO : 25 mg/mL (33.16 mM; Need ultrasonic)
Indhold og opbevaring -20°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Forsendelsestilstand Room temperature
Mål HBV
CAS 146368-14-1
Synonym Cy5 NHS Ester ∣ Sulfo-Cyanine5 Succinimidyl Ester
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.