CAS RN 140-64-7
CAS RN 140-64-7
IUPAC Navn:
bis(2-hydroxyethane-1-sulfonic acid); 4-{[5-(4-carbamimidoylphenoxy)pentyl]oxy}benzene-1-carboximidamide
Synonymer:
bis(isethionic acid); pentamidine
Molekylvægt (g/mol):
592.68
Molekylformel:
C23H36N4O10S2
InChi Key:
YBVNFKZSMZGRAD-UHFFFAOYSA-N
SMILES:
OCCS(O)(=O)=O.OCCS(O)(=O)=O.NC(=N)C1=CC=C(OCCCCCOC2=CC=C(C=C2)C(N)=N)C=C1
1
–
4
af
4
Resultater
1
Kampagner er tilgængelige
Thermo Scientific Chemicals Pentamidine isethionatsalt, 97%
Thermo Scientific Chemicals Pentamidine isethionatsalt, 97%
2
Pentamidine Diisetionate, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
3
Pentamidine Diisetionate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4
Pentamidine isethionate, MedChemExpress
MedChemExpress Pentamidine isethionate (MP-601205 isethionate) is an antimicrobial agent and interferes with DNA biosynthetics.