CAS RN 129938-20-1
CAS RN 129938-20-1
IUPAC Navn:
hydrogen dimethyl[(1S)-3-(naphthalen-1-yloxy)-1-phenylpropyl]amine chloride
Synonymer:
dapoxetine
Molekylvægt (g/mol):
341.88
Molekylformel:
C21H24ClNO
InChi Key:
IHWDIQRWYNMKFM-BDQAORGHSA-N
SMILES:
[H+].[Cl-].CN(C)[C@@H](CCOC1=C2C=CC=CC2=CC=C1)C1=CC=CC=C1
1
–
2
af
2
Resultater
1
Dapoxetine Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.