CAS RN 103597-45-1
CAS RN 103597-45-1
IUPAC Navn:
2-(2H-1,2,3-benzotriazol-2-yl)-6-{[3-(2H-1,2,3-benzotriazol-2-yl)-2-hydroxy-5-(2,4,4-trimethylpentan-2-yl)phenyl]methyl}-4-(2,4,4-trimethylpentan-2-yl)phenol
Synonymer:
bisoctrizole
Molekylvægt (g/mol):
658.89
Molekylformel:
C41H50N6O2
InChi Key:
FQUNFJULCYSSOP-UHFFFAOYSA-N
SMILES:
CC(C)(C)CC(C)(C)C1=CC(N2N=C3C=CC=CC3=N2)=C(O)C(CC2=C(O)C(=CC(=C2)C(C)(C)CC(C)(C)C)N2N=C3C=CC=CC3=N2)=C1
1
–
2
af
2
Resultater
1
Bisoctrizole, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.