CAS RN 1020719-68-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
6
af
6
Resultater
1
(R,S)-N-Nitroso Anabasine-d4 (0.1 mg/mL in Methanol), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Mixture of (R,S)-N-Nitroso Anabasine-d4, rac N'-Nitrosonornicotine-d4, 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanone-d4 and (S)-N-Nitroso Anatabine-d4 (1.0mg/mL in Acetonitrile), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(R,S)-N-Nitroso Anabasine-d4, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
(R,S)-N-Nitroso Anatabine-2,4,5,6-d4 (10 ug/ml in Acetonitrile), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
6
(R,S)-N-Nitroso Anabasine-d4, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.