CAS RN 97978-07-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
Benzo[b]thien-2-yl Ketone (Zileuton Impurity), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Zileuton USP Related Compound B, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Benzo[b]thien-2-yl Ketone (Zileuton Impurity), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Benzo[b]thien-2-yl Ketone (Zileuton Impurity), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.