CAS RN 93951-97-4
IUPAC Navn:
acenaphthylene
Synonymer:
acenaphthylene cyclopenta(de)naphthalene acenaphthalene
Molekylvægt (g/mol):
152.2
Molekylformel:
C12H8
InChi Key:
HXGDTGSAIMULJN-UHFFFAOYSA-N
SMILES:
C1=CC2=C3C1=CC=CC3=CC=C2
1
–
3
af
3
Resultater
1
Acenaphthylene-d8, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Acenaphthylene-d8, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.