CAS RN 896720-20-0
IUPAC Navn:
4-{2-[(2-chloro-4-fluorophenyl)amino]-5-methylpyrimidin-4-yl}-N-[1-(3-chlorophenyl)-2-hydroxyethyl]-1H-pyrrole-2-carboxamide
Synonymer:
4-{2-[(2-chloro-4-fluorophenyl)amino]-5-methylpyrimidin-4-yl}-N-[1-(3-chlorophenyl)-2-hydroxyethyl]-1H-pyrrole-2-carboxamide
Molekylvægt (g/mol):
500.36
Molekylformel:
C24H20Cl2FN5O2
InChi Key:
WUTVMXLIGHTZJC-UHFFFAOYNA-N
SMILES:
CC1=CN=C(NC2=C(Cl)C=C(F)C=C2)N=C1C1=CNC(=C1)C(=O)NC(CO)C1=CC(Cl)=CC=C1
1
–
2
af
2
Resultater
2
VX-11e, MedChemExpress
MedChemExpress VX-11e is a potent, selective, and orally bioavailable inhibitor of ERK with Ki < 2 nM.