CAS RN 89391-50-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Imirestat, MedChemExpress
MedChemExpress Imirestat (AL 1576) is an aldose reductase inhibitor, used for the treatment of diabetes.