CAS RN 892489-52-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
AZD3988, MedChemExpress
MedChemExpress AZD3988 is a diacylglycerol acyl transferase-1 (DGAT-1) inhibitor with IC50s of 6, 5, 11 nM for human, rat, and mouse DGAT-1, respectively.