CAS RN 89059-37-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Nitrobenzene-13C6, TRC
CAS: 89059-37-0 Molekylær formel: 13C6 H5 N O2 Molekylvægt (g/mol): 129.07 Synonym: Benzene-1,2,3,4,5,6-13C6, 1-nitro-,Benzene-13C6, nitro- (9CI),1-Nitrobenzene-1,2,3,4,5,6-13C6,Nitrobenzene-13C6,Nitrobenzene 13C6 IUPAC navn: nitro(1,2,3,4,5,6-^{13}C_{6})cyclohexatriene SMIL: [O-][N+](=O)[13c]1[13cH][13cH][13cH][13cH][13cH]1
2
Nitrobenzene 13C6 100 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality