Få mere at vide
Nitrobenzene-13C6, TRC
Artikelnummer. 30473752
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30473752 10 mg
30470014 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30473752 Leverandør Toronto Research Chemicals Leverandørnr. TRCN49300310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 89059-37-0
Molekylær formel 13C6 H5 N O2
Molekylvægt (g/mol) 129.07
Synonym Benzene-1,2,3,4,5,6-13C6, 1-nitro-, Benzene-13C6, nitro- (9CI), 1-Nitrobenzene-1,2,3,4,5,6-13C6, Nitrobenzene-13C6, Nitrobenzene 13C6
IUPAC navn nitro(1,2,3,4,5,6-^{13}C_{6})cyclohexatriene
SMIL [O-][N+](=O)[13c]1[13cH][13cH][13cH][13cH][13cH]1
Tekniske data
Kemisk navn eller materiale Nitrobenzene 13C6
Mængde 10 mg
InChI formel InChI=1S/C6H5NO2/c8-7(9)6-4-2-1-3-5-6/h1-5H/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vægt 129.0522