CAS RN 887973-21-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(Z)-Fluoxastrobin 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
(Z)-Fluoxastrobin, TRC
CAS: 887973-21-9 Molekylær formel: C21 H16 Cl F N4 O5 Molekylvægt (g/mol): 458.83 Synonym: Methanone, [2-[[6-(2-chlorophenoxy)-5-fluoro-4-pyrimidinyl]oxy]phenyl](5,6-dihydro-1,4,2-dioxazin-3-yl)-, O-methyloxime, (1Z)-,(Z)-Fluoxastrobin IUPAC navn: (Z)-1-[2-[6-(2-chlorophenoxy)-5-fluoropyrimidin-4-yl]oxyphenyl]-1-(5,6-dihydro-1,4,2-dioxazin-3-yl)-N-methoxymethanimine SMIL: CO\N=C(/C1=NOCCO1)\c2ccccc2Oc3ncnc(Oc4ccccc4Cl)c3F