CAS RN 885434-70-8
IUPAC Navn:
2,4-dimethyl-1-(2,4,6-trimethylbenzenesulfonyl)-1H-pyrrole
Synonymer:
2,4-dimethyl-1-(2,4,6-trimethylbenzenesulfonyl)pyrrole
Molekylvægt (g/mol):
277.38
Molekylformel:
C15H19NO2S
InChi Key:
AFZWZVLPIMHLSE-UHFFFAOYSA-N
SMILES:
CC1=CN(C(C)=C1)S(=O)(=O)C1=C(C)C=C(C)C=C1C
1
–
2
af
2
Resultater
1
Tocris Bioscience™ HJC 0350
CAS: 885434-70-8 Molekylær formel: C15H19NO2S
2
HJC0350, MedChemExpress
MedChemExpress HJC0350 is a potent and specific EPAC2 antagonist with an IC50 of 0.3 μM.