CAS RN 860033-06-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
L 888607, MedChemExpress
MedChemExpress L 888607 is a potent, and selective CRTH2 (also known as DP2) agonist with a Ki of 0.8 nM.