CAS RN 853792-81-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
1-Propyl-1H-imidazo[4,5-c]quinolin-4-amine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Desmethyl-N-propyl Imiquimod, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.