CAS RN 85-70-1
IUPAC Navn:
butyl 2-[(2-butoxy-2-oxoethoxy)carbonyl]benzoate
Synonymer:
butyl phthalyl butyl glycolate
Molekylvægt (g/mol):
336.38
Molekylformel:
C18H24O6
InChi Key:
GOJCZVPJCKEBQV-UHFFFAOYSA-N
SMILES:
CCCCOC(=O)COC(=O)C1=CC=CC=C1C(=O)OCCCC
1
–
3
af
3
Resultater
1
Butyl Phthalyl Butyl Glycolate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Butyl Phthalyl Butyl Glycolate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Butyl Phthalyl Butyl Glycolate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.