Få mere at vide
Butyl Phthalyl Butyl Glycolate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Phthalic acid, 2-butoxy-2-oxoethyl butyl ester |
| CAS | 85-70-1 |
| Molekylær formel | C18 H24 O6 |
| Mængde | 500 mg |
| Synonym | 1,2-Benzenedicarboxylic acid, 2-butoxy-2-oxoethyl butyl ester (9CI), Phthalic acid, butyl ester, ester with butyl glycolate (6CI,7CI,8CI), Glycolic acid, butyl ester, butyl phthalate (8CI), 2-Butoxy-2-oxoethyl butyl phthalate, 2-O-(2-Butoxy-2-oxoethyl) 1-O-butyl benzene-1,2-dicarboxylate, Butoxycarbonylmethyl butyl phthalate, Butyl 2-butoxy-2-exoethyl phthalate, Butyl carbobutoxymethyl phthalate, Butyl glycolyl butyl phthalate, Butyl phthalate butyl glycolate, Butyl phthalyl butyl glycolate, Dibutyl o-carboxybenzoyloxyacetate, Morflex 190, Reomol 4PG, Santicizer B 16, Phthalic Acid, 2-Butoxy-2-oxoethyl Butyl Ester |
| InChI formel | InChI=1S/C18H24O6/c1-3-5-11-22-16(19)13-24-18(21)15-10-8-7-9-14(15)17(20)23-12-6-4-2/h7-10H,3-6,11-13H2,1-2H3 |
| SMIL | CCCCOC(=O)COC(=O)c1ccccc1C(=O)OCCCC |
| IUPAC navn | 2-O-(2-butoxy-2-oxoethyl) 1-O-butyl benzene-1,2-dicarboxylate |
| Molekylvægt (g/mol) | 336.3796 |
| Formel vægt | 336.1573 |