CAS RN 849727-62-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
3-(2-Chloroethyl)-9-hydroxy-2-methyl-4H-Pyrido[1,2-a]pyrimidin-4-one Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-(2-Chloroethyl)-9-hydroxy-2-methyl-4H-Pyrido[1,2-a]pyrimidin-4-one Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.