CAS RN 84632-65-5
IUPAC Navn:
3,6-bis(4-chlorophenyl)-1H,2H,4H,5H-pyrrolo[3,4-c]pyrrole-1,4-dione
Synonymer:
3,6-bis(4-chlorophenyl)-2H,5H-pyrrolo[3,4-c]pyrrole-1,4-dione
Molekylvægt (g/mol):
357.19
Molekylformel:
C18H10Cl2N2O2
InChi Key:
JNNHVXMCVRYTTN-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C=C1)C1=C2C(=O)NC(=C2C(=O)N1)C1=CC=C(Cl)C=C1
1
–
3
af
3
Resultater
1
3,6-Bis(4-chlorophenyl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3,6-Bis(4-chlorophenyl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3,6-Bis(4-chlorophenyl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.