missing translation for 'onlineSavingsMsg'
Få mere at vide

3,6-Bis(4-chlorophenyl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione (Technical Grade), TRC

Artikelnummer. 30366803
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366803 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30366803 Leverandør Toronto Research Chemicals Leverandørnr. TRCC37543350MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 3,6-Bis(4-chlorophenyl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione
CAS 84632-65-5
Molekylær formel C18H10Cl2N2O2
Mængde 50 mg
Synonym 3,6-Bis(4-chlorophenyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione, 1,4-Bis(4-chlorophenyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-3,6-dione, 1,4-Diketo-3,6-bis(4-chlorophenyl)pyrrolo[3,4-c]pyrrole, 1,4-Diketo-3,6-di(4-chlorophenyl)pyrrolo[3,4-c]pyrrole, 3,6-Bis(4-chlorophenyl)-1,4-diketopyrrolo[3,4-c]pyrrole, 3,6-Bis(4-chlorophenyl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione, BK-CF, C.I. 56110, C.I. PR 254, C.I. Pigment Red 254, CFP-FF 449R, Chromofine Red 6156, Chromofine Red 6156EC, Cinilex DPP Red SR 1C, Cinilex DPP Red SR 2P, Cinilex DPP Red ST, Cromophtal DPP Red, Cromophtal DPP Red BO, Cromophtal DPP Red BOC, Cromophtal DPP Red BP, Cromophtal DPP Red BP-P, Cromophtal DPP Red RP, Cromophtal Red 2028, Cromophtal Red 2030, Cromophtal Red B-CF, Cromophtal Red BP, Cromophtal Red DPP, DDP Red SR 1C, DPP Red BO, DPP Red BOC, DPP Red BTR, DPP Red SR 2P, DPP Red ST, DPP-BO, Eupolen PE Red 34-3001, Fastogen Super Red 254-226-0200, Fuji Fast Red 8800, Fuji Red 8800, Hostaperm Red D 2B-COF-LV3781, Hostaperm Red D 2B-COFO1, Hostaperm Red D 3G70, Hostaperm Red D 3G70EDS, Hostaperm Red D 3G70EDW, Irgaphor BK-CF, Irgaphor BT-CF, Irgaphor Red 2030, Irgaphor Red B-CF, Irgaphor Red BK-CF, Irgaphor Red BT-CF, Irgaphor Red S 3610CF, Irgazin DPP Red 2030, Irgazin DPP Red BO, Irgazin DPP Red BOX, Irgazin DPP Red BP, Irgazin DPP Red BTR, Irgazin RED L 3837HD, Irgazin Red 2030, Irgazin Red 3630, Irgazin Red BO, Irgazin Red BOX, Irgazin Red D 3656HD, Irgazin Red DPP-BO, Irgazin Red K 3842, Irgazin Red L 3660HD, Irgazin Red L 3670HD, Lionogen Red LX 8318, Luconyl NG Red 3860, Microlen DPP RED BP, Microlith DPP Red B-K, PR 254, PV Fast Red D 3G, Pigment Red 254, RT 107, Red BT-CF, Versal Red DP 3G, Vynamon Red 325401, Xfast Red 3860, Xianlite Red EI 3333
InChI formel InChI=1S/C18H10Cl2N2O2/c19-11-5-1-9(2-6-11)15-13-14(18(24)21-15)16(22-17(13)23)10-3-7-12(20)8-4-10/h1-8,21,24H
SMIL ClC1=CC=C(C2=C3C(C(N2)=O)=C(NC3=O)C4=CC=C(Cl)C=C4)C=C1
IUPAC navn 1,4-bis(4-chlorophenyl)-3-hydroxy-2H-pyrrolo[3,4-c]pyrrol-6-one
Molekylvægt (g/mol) 357.19
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.