CAS RN 84611-22-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
2-Chloro-N-(2-oxotetrahydrothiophen-3-yl)acetamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
N-Chloroacetyl DL-Homocysteine Thiolactone, TRC
CAS: 84611-22-3 Molekylær formel: C6 H8 Cl N O2 S Molekylvægt (g/mol): 193.65 Synonym: 2-Chloro-N-(2-oxotetrahydrothiophen-3-yl)acetamide IUPAC navn: 2-chloro-N-(2-oxothiolan-3-yl)acetamide SMIL: ClCC(=O)NC1CCSC1=O