Få mere at vide
N-Chloroacetyl DL-Homocysteine Thiolactone, TRC
Artikelnummer. 17962975
Skift visning
Mængde:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17962975 50 g
17924985 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17962975 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36348050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 84611-22-3
Molekylær formel C6 H8 Cl N O2 S
Molekylvægt (g/mol) 193.65
Synonym 2-Chloro-N-(2-oxotetrahydrothiophen-3-yl)acetamide
IUPAC navn 2-chloro-N-(2-oxothiolan-3-yl)acetamide
SMIL ClCC(=O)NC1CCSC1=O
Tekniske data
Kemisk navn eller materiale 2-Chloro-N-(2-oxotetrahydrothiophen-3-yl)acetamide
Renhedsgrad noter HPLC
InChI formel InChI=1S/C6H8ClNO2S/c7-3-5(9)8-4-1-2-11-6(4)10/h4H,1-3H2,(H,8,9)
Mængde 50 g
Formel vægt 192.9964
Procent renhed >95%