CAS RN 83881-08-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
17Alpha-Hydroxy-21-chloro-9Beta,11Beta-epoxy-16Alpha-methylpregna-1,4-diene-3,20-dione, TRC
CAS: 83881-08-7 Molekylær formel: C22 H27 Cl O4 Molekylvægt (g/mol): 390.9 Synonym: Mometasone Furoate Impurity Q IUPAC navn: (1S,2S,10S,11S,13R,14R,15S,17S)-14-(2-chloroacetyl)-14-hydroxy-2,13,15-trimethyl-18-oxapentacyclo[8.8.0.01,17.02,7.011,15]octadeca-3,6-dien-5-one SMIL: C[C@@H]1C[C@H]2[C@@H]3CC=C4CC(=O)C=C[C@]4(C)[C@@]35O[C@H]5C[C@]2(C)[C@@]1(O)C(=O)CCl
2
21-Chloro-9,11beta-epoxy-17-hydroxy-16alpha-methyl-9beta-pregna-1,4-diene-3,20-dione, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.