CAS RN 80879-63-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Emiglitate, MedChemExpress
MedChemExpress Emiglitate (BAY o 1248) is a potent, selective and competitive inhibitor of α-glucoside hydrolase.