CAS RN 794477-75-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
N2-[1-Deoxy-2,3:4,5-bis-O-(1-methylethylidene)-Beta-D-fructopyranos-1-yl]-L-asparagine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N2-[1-Deoxy-2,3:4,5-bis-O-(1-methylethylidene)-Beta-D-fructopyranos-1-yl]-L-asparagine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.