CAS RN 79-46-9
IUPAC Navn:
2-nitropropane
Synonymer:
β-nitropropane
Molekylvægt (g/mol):
89.09
Molekylformel:
C3H7NO2
InChi Key:
FGLBSLMDCBOPQK-UHFFFAOYSA-N
SMILES:
CC(C)[N+]([O-])=O
1
–
4
af
4
Resultater
1
2-Nitropropane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Nitropropane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Nitropropane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.