Få mere at vide
2-Nitropropane, SPEX CertiPrep™
Artikelnummer. 12939314 Shop alle SPEX Certiprep produkter
Skift visning
Mængde:
1 mL
Pakningsstørrelse:
1ml
Artikelnummer. Mængde unitSize
12939314 1 mL 1ml
1 options
Artikelnummer. 12939314 Leverandør SPEX Certiprep Leverandørnr. S2732
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Indeholder
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
2-Nitropropane79-46-91000μg/mL
Certificeringer
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Overholdelse
GHS/OSHA Complaint
Tekniske data
Kemisk navn eller materiale 2-Nitropropane
Emballage 2mL Amber Prescored Glass Ampule
CAS 79-46-9, 67-56-1
CAS min % 0.1, 99.9
Koncentration eller sammensætningsnoter Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molekylær formel C3H7NO2
Til brug med (udstyr) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI nøgle FGLBSLMDCBOPQK-UHFFFAOYSA-N
SMIL CC(C)[N+](=O)[O-]
IUPAC navn 2-nitropropane
Opløsningsmiddel eller Matrix Methanol-P&T
Molekylvægt (g/mol) 89.094
PubChem CID 398
Mængde 1 mL
ChEBI CHEBI:16037
Vis mere Vis mindre
Product intended for laboratory use only