CAS RN 7682-18-0
IUPAC Navn:
methyl 2-aminobutanoate hydrochloride
Synonymer:
methyl 2-aminobutanoate hydrochloride
Molekylvægt (g/mol):
153.61
Molekylformel:
C5H12ClNO2
InChi Key:
AHAQQEGUPULIOZ-UHFFFAOYNA-N
SMILES:
Cl.CCC(N)C(=O)OC
1
–
2
af
2
Resultater
1
DL-2-Aminobutyric Acid Methyl Ester Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
DL-2-Aminobutyric Acid Methyl Ester Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.