CAS RN 75659-08-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(R,R)-Labetalol Hydrochloride, TRC
CAS: 75659-08-4 Molekylær formel: C19 H24 N2 O3 . Cl H Molekylvægt (g/mol): 364.87 Synonym: Benzamide, 2-hydroxy-5-[(1R)-1-hydroxy-2-[[(1R)-1-methyl-3-phenylpropyl]amino]ethyl]-, hydrochloride (1:1),Benzamide, 2-hydroxy-5-[(1R)-1-hydroxy-2-[[(1R)-1-methyl-3-phenylpropyl]amino]ethyl]-, monohydrochloride (9CI),Benzamide, 2-hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]-, monohydrochloride, [R-(R*,R*)]-,(R,R)-Labetalol hydrochloride,Dilevalol hydrochloride,Sch 19927 HCl IUPAC navn: 2-hydroxy-5-[(1R)-1-hydroxy-2-[[(2R)-4-phenylbutan-2-yl]amino]ethyl]benzamide;hydrochloride SMIL: Cl.C[C@H](CCc1ccccc1)NC[C@H](O)c2ccc(O)c(c2)C(=O)N