CAS RN 754240-09-0
IUPAC Navn:
2-[(morpholin-4-yl)methyl]-5-[(5-{[7-(trifluoromethyl)quinolin-4-yl]sulfanyl}pentyl)oxy]-4H-pyran-4-one dihydrochloride
Synonymer:
2-(morpholin-4-ylmethyl)-5-[(5-{[7-(trifluoromethyl)quinolin-4-yl]sulfanyl}pentyl)oxy]pyran-4-one dihydrochloride
Molekylvægt (g/mol):
581.47
Molekylformel:
C25H29Cl2F3N2O4S
InChi Key:
LSECOAJFCKFQJG-UHFFFAOYSA-N
SMILES:
Cl.Cl.FC(F)(F)C1=CC=C2C(SCCCCCOC3=COC(CN4CCOCC4)=CC3=O)=CC=NC2=C1
1
–
3
af
3
Resultater
1
Tocris Bioscience™ EHT 1864
CAS: 754240-09-0 Molekylær formel: C25H29Cl2F3N2O4S
2
EHT 1864, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3