CAS RN 751455-55-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Resorcinol-d4, TRC
CAS: 751455-55-7 Molekylær formel: C6 2H4 H2 O2 Molekylvægt (g/mol): 114.14 Synonym: 1,3-Benzene-2,4,5,6-d4-diol (9CI),1,3-Benzenediol-d4,Resorcinol-d4,Resorcinol-2,4,5,6-D4,1,3-Dihydroxybenzene-2,4,5,6-d4 IUPAC navn: 2,4,5,6-tetradeuteriobenzene-1,3-diol SMIL: [2H]c1c([2H])c(O)c([2H])c(O)c1[2H]
2
Resorcinol-d4, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Resorcinol-d4, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.