Få mere at vide
Resorcinol-d4, TRC
Artikelnummer. 30410975
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30410975 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30410975 Leverandør Toronto Research Chemicals Leverandørnr. TRCR144704100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Resorcinol-d4
CAS 751455-55-7
Molekylær formel C6 2H4 H2 O2
Mængde 100 mg
Synonym 1,3-Benzene-2,4,5,6-d4-diol (9CI), 1,3-Benzenediol-d4, Resorcinol-d4, Resorcinol-2,4,5,6-D4, 1,3-Dihydroxybenzene-2,4,5,6-d4
InChI formel InChI=1S/C6H6O2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(O)c([2H])c(O)c1[2H]
IUPAC navn 2,4,5,6-tetradeuteriobenzene-1,3-diol
Molekylvægt (g/mol) 114.14
Formel vægt 114.0619