CAS RN 745833-23-2
IUPAC Navn:
2-(2,4-difluorophenyl)-6-[1-(2,6-difluorophenyl)carbamoylamino]pyridine-3-carboxamide
Synonymer:
2-(2,4-difluorophenyl)-6-[1-(2,6-difluorophenyl)carbamoylamino]pyridine-3-carboxamide
Molekylvægt (g/mol):
404.33
Molekylformel:
C19H12F4N4O2
InChi Key:
FYSRKRZDBHOFAY-UHFFFAOYSA-N
SMILES:
NC(=O)N(C1=NC(=C(C=C1)C(N)=O)C1=C(F)C=C(F)C=C1)C1=C(F)C=CC=C1F
1
–
1
af
1
Resultater
1
VX-702, MedChemExpress
MedChemExpress VX-702 is a highly selective inhibitor of p38α MAPK, 14-fold higher potency against the p38α versus p38β.