CAS RN 71901-21-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
5
af
5
Resultater
1
MedChemExpress N-Formyl-Nle-Leu-Phe-Nle-Tyr-Lys
MedChemExpress N-Formyl-Nle-Leu-Phe-Nle-Tyr-Lys TFA (For-Nle-Leu-Phe-Nle-Tyr-Lys-OH TFA) is a formyl peptide receptor (FPR) agonist.
2
MedChemExpress N-Formyl-Nle-Leu-Phe-Nle-Tyr-Lys
MedChemExpress N-Formyl-Nle-Leu-Phe-Nle-Tyr-Lys TFA (For-Nle-Leu-Phe-Nle-Tyr-Lys-OH TFA) is a formyl peptide receptor (FPR) agonist.
3
MedChemExpress N-Formyl-Nle-Leu-Phe-Nle-Tyr-Lys
MedChemExpress N-Formyl-Nle-Leu-Phe-Nle-Tyr-Lys TFA (For-Nle-Leu-Phe-Nle-Tyr-Lys-OH TFA) is a formyl peptide receptor (FPR) agonist.
4
MedChemExpress N-Formyl-Nle-Leu-Phe-Nle-Tyr-Lys
MedChemExpress N-Formyl-Nle-Leu-Phe-Nle-Tyr-Lys TFA (For-Nle-Leu-Phe-Nle-Tyr-Lys-OH TFA) is a formyl peptide receptor (FPR) agonist.
5
MedChemExpress N-Formyl-Nle-Leu-Phe-Nle-Tyr-Lys
MedChemExpress N-Formyl-Nle-Leu-Phe-Nle-Tyr-Lys TFA (For-Nle-Leu-Phe-Nle-Tyr-Lys-OH TFA) is a formyl peptide receptor (FPR) agonist.