CAS RN 7147-77-5
IUPAC Navn:
5-(4-nitrophenyl)furan-2-carbaldehyde
Synonymer:
5-(4-nitrophenyl)furan-2-carbaldehyde
Molekylvægt (g/mol):
217.18
Molekylformel:
C11H7NO4
InChi Key:
RTSOJVJDKNKNFU-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(C=C1)C1=CC=C(O1)C=O
1
–
2
af
2
Resultater
1
5-(4-Nitrophenyl)furfural, TRC
CAS: 7147-77-5 Molekylær formel: C11 H7 N O4 Molekylvægt (g/mol): 217.18 Synonym: 5-(4-Nitrophenyl)furan-2-carbaldehyde,5-(4-Nitrophenyl)-2-furancarboxyaldehyde IUPAC navn: 5-(4-nitrophenyl)furan-2-carbaldehyde SMIL: [O-][N+](=O)c1ccc(cc1)c2oc(C=O)cc2
2
Dantrolene USP Related Compound C, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.