Få mere at vide
Dantrolene USP Related Compound C, Mikromol™
Artikelnummer. 30464214
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30464214 4 x 25 mg
30052655 25 mg
17963196 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30464214 Leverandør Mikromol Leverandørnr. MM0905.0100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Dantrolene USP Related Compound C, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-(4-nitrophenyl)-2-furancarboxyaldehyde
CAS 7147-77-5
Analyt- eller komponentnavne 5-(4-nitrophenyl)-2-furancarboxyaldehyde
Forsendelsestilstand Room Temperature
Molekylær formel C11 H7 N O4
Certificeringer/compliance ISO 17025
Synonym 5-(4-Nitrophenyl)furan-2-carbaldehyde, (5-(4-nitrophenyl)-2-furancarboxyaldehyde, Dantrolene Related Compound C
InChI formel InChI=1S/C11H7NO4/c13-7-10-5-6-11(16-10)8-1-3-9(4-2-8)12(14)15/h1-7H
Løsningstype Neat
SMIL [O-][N+](=O)c1ccc(cc1)c2oc(C=O)cc2
IUPAC navn 5-(4-nitrophenyl)furan-2-carbaldehyde
Molekylvægt (g/mol) 217.18
Mængde 4 x 25 mg
Formel vægt 217.0375 g/mol