CAS RN 70484-01-4
IUPAC Navn:
4-bromobenzene-1,2-dicarbonitrile
Synonymer:
4-bromobenzene-1,2-dicarbonitrile
Molekylvægt (g/mol):
207.03
Molekylformel:
C8H3BrN2
InChi Key:
ARAONWRSVQOHIT-UHFFFAOYSA-N
SMILES:
BrC1=CC(C#N)=C(C=C1)C#N
1
–
3
af
3
Resultater
1
4-Bromophthalonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Bromophthalonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Bromophthalonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.