CAS RN 700878-19-9
IUPAC Navn:
2-({4-[(2-fluorophenyl)methyl]piperidin-1-yl}methyl)-1H-1,3-benzodiazol-6-ol hydrochloride
Synonymer:
2-({4-[(2-fluorophenyl)methyl]piperidin-1-yl}methyl)-3H-1,3-benzodiazol-5-ol hydrochloride
Molekylvægt (g/mol):
375.87
Molekylformel:
C20H23ClFN3O
InChi Key:
IJHXPTYEGXYLBZ-UHFFFAOYSA-N
SMILES:
Cl.OC1=CC=C2N=C(CN3CCC(CC4=CC=CC=C4F)CC3)NC2=C1
1
–
1
af
1
Resultater
1
NMDA-IN-1, MedChemExpress
MedChemExpress NMDA-IN-1 is a potent and NR2B-selective NMDA antagonist with Ki of 0.85 nM; NR2B Ca2+ influx IC50 is 9.7 nM; no activities on NR2A, NR2C, NR2D, hERG-channel and α1-adrenergic receptor.