CAS RN 698-87-3
IUPAC Navn:
1-phenylpropan-2-ol
Synonymer:
1-phenyl-2-propanol
Molekylvægt (g/mol):
136.19
Molekylformel:
C9H12O
InChi Key:
WYTRYIUQUDTGSX-UHFFFAOYNA-N
SMILES:
CC(O)CC1=CC=CC=C1
1
–
4
af
4
Resultater
1
Methylbenzeneethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-Phenyl-2-propanol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Methylbenzeneethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Methylbenzeneethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.