missing translation for 'onlineSavingsMsg'
Få mere at vide

1-Phenyl-2-propanol, Mikromol™

Product Code. 30049919
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30049919 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049919 Supplier Mikromol Supplier No. MM0741.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-Phenyl-2-propanol
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 1-Phenyl-2-propanol
CAS 698-87-3
Analyt- eller komponentnavne 1-Phenyl-2-propanol
Forsendelsestilstand Room Temperature
Molekylær formel C9 H12 O
Certificeringer/compliance ISO 17025
Synonym alpha-Methylbenzeneethanol, alpha-Methylphenethyl alcohol, (+/-)-1-Phenylpropan-2-ol, (+/-)-alpha-Methylbenzeneethanol, (+/-)-alpha-Methylphenethyl alcohol, 1-Phenyl-2-propanol, 2-Hydroxy-1-phenylpropane, 1-Phenyl-2-propanol, 3-Phenyl-2-hydroxypropane, 3-Phenyl-2-propanol, Benzyl methyl carbinol, Benzylethyl alcohol, NSC 53553, alpha-Methylbenzeneethanol, alpha-Methylphenethyl alcohol, Dextroamphetamine Related Compound A, Amfetamine Sulfate Impurity A, Dexamfetamine Sulfate Impurity A, Amfetamine Impurity A, Dexamfetamine Impurity A
InChI formel InChI=1S/C9H12O/c1-8(10)7-9-5-3-2-4-6-9/h2-6,8,10H,7H2,1H3
SMIL CC(O)Cc1ccccc1
IUPAC navn 1-phenylpropan-2-ol
Molekylvægt (g/mol) 136.19
Mængde 100 mg
Formel vægt 136.0888 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.