CAS RN 688805-40-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
MedChemExpress HEX3
MedChemExpress HEX3 is a fragment of the adenoviral hexon. Hexon is the major capsid protein of adenovirion and is comprised of three identical polypeptide chains.
2
MedChemExpress HEX3
MedChemExpress HEX3 is a fragment of the adenoviral hexon. Hexon is the major capsid protein of adenovirion and is comprised of three identical polypeptide chains.
3
MedChemExpress HEX3
MedChemExpress HEX3 is a fragment of the adenoviral hexon. Hexon is the major capsid protein of adenovirion and is comprised of three identical polypeptide chains.