CAS RN 67700-30-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Furaprofen, MedChemExpress
MedChemExpress Furaprofen (R803) is an effective HCV replication inhibitor. Furaprofen (R803) is substantially more potent against genotype 1a and 1b replicons (EC50, ∼30 nM) than against the genotype 2a replicon (EC50, ∼1,000 nM).