CAS RN 67006-34-2
IUPAC Navn:
2-[2-hydroxy-3-(2-hydroxyethanesulfonyl)propanesulfonyl]ethan-1-ol
Synonymer:
2-[2-hydroxy-3-(2-hydroxyethanesulfonyl)propanesulfonyl]ethanol
Molekylvægt (g/mol):
276.32
Molekylformel:
C7H16O7S2
InChi Key:
CVFXDAXIZHJZAH-UHFFFAOYSA-N
SMILES:
OCCS(=O)(=O)CC(O)CS(=O)(=O)CCO
1
–
3
af
3
Resultater
1
2,2'-(2-Hydroxypropane-1,3-diyldisulfonyl)diethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,2'-(2-Hydroxypropane-1,3-diyldisulfonyl)diethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,2'-(2-Hydroxypropane-1,3-diyldisulfonyl)diethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.