CAS RN 666174-80-7
IUPAC Navn:
2,5-dihydroxybenzoic acid; butan-1-amine
Synonymer:
2,5-dihydroxybenzoic acid; butylamine
Molekylvægt (g/mol):
227.26
Molekylformel:
C11H17NO4
InChi Key:
OGLUKKYDTRVMHP-UHFFFAOYSA-N
SMILES:
CCCCN.OC(=O)C1=C(O)C=CC(O)=C1
1
–
3
af
3
Resultater
1
2,5-Dihydroxybenzoic Acid Butylamine Salt [Matrix for MALDI-TOF/MS], TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,5-Dihydroxybenzoic Acid Butylamine Salt [Matrix for MALDI-TOF/MS], TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,5-Dihydroxybenzoic Acid Butylamine Salt [Matrix for MALDI-TOF/MS], TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.