CAS RN 65447-77-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Poly(4-hydroxy-2,2,6,6-tetramethyl-1-piperidineethanol-alt-1,4-butanedioic Acid) (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Poly(4-hydroxy-2,2,6,6-tetramethyl-1-piperidineethanol-alt-1,4-butanedioic Acid), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Poly(4-hydroxy-2,2,6,6-tetramethyl-1-piperidineethanol-alt-1,4-butanedioic Acid) (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.