CAS RN 654080-03-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
SK-216, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Tocris Bioscience™ SK 216
Plasminogen activator inhibitor-1 (PAI-1) inhibitor
3
SK-216, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
SK-216, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.